| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011497.hg.1 |
ENST00000515414.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011497.hg.1 |
ENST00000515414.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011497.hg.1 |
ENST00000515414.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011497.hg.1 |
ENST00000515414.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011497.hg.1 |
ENST00000515414.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011497.hg.1 |
NR_126420.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011497.hg.1 |
NR_126420.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011497.hg.1 |
NR_126420.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011497.hg.1 |
NR_126420.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011497.hg.1 |
NR_126420.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011497.hg.1 |
NR_126420.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011497.hg.1 |
ENST00000515414.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011497.hg.1 |
ENST00000515414.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011497.hg.1 |
ENST00000515414.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011497.hg.1 |
ENST00000515414.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011497.hg.1 |
ENST00000515414.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011497.hg.1 |
NR_126420.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011497.hg.1 |
NR_126420.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011497.hg.1 |
NR_126420.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011497.hg.1 |
NR_126420.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011497.hg.1 |
NR_126420.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011497.hg.1 |
NR_126420.1 |
Refseq |