Assay Details
| Gene Symbol: SALRNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126481.1 | 1 | 619 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929151 | LOC101929151 | -28.9kb | Anti-sense | Chr. 15 - 70570958 - 70586606 on NCBI Build GRCh38 | |
| Downstream | senescence associated long non-coding RNA 2 | SALRNA2 |
SAL-RNA2, XLOC_025931 |
18.7kb | Sense | Chr. 15 - 70635249 - 70637314 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SALRNA3
Genomic Map