Assay Details
Gene Symbol: C20orf197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_126504.1 | 1 - 2 | 84 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cadherin 26 | CDH26 | VR20 | -20.3kb | Sense | Chr. 20 - 59957764 - 60035629 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC729296 | LOC729296 | 29.8kb | Sense | Chr. 20 - 60087686 - 60101387 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
