Assay Details
Gene Symbol: OR7E91P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002185.3 | 2 - 3 | 220 | 88 | ||
GenBank mRNA | BC014374.1 | - | 2 - 3 | 223 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | testis expressed 261 | TEX261 | TEG-261 | -29.4kb | Anti-sense | Chr. 2 - 70985938 - 70994871 on NCBI Build GRCh38 |
Downstream | olfactory receptor family 7 subfamily E member 62 pseudogene | OR7E62P |
OR2-4, OR2-52, OR2-53, OR2-75, OR7E62, OR7E63, OR7E63P, OR7E64, OR7E64P, OR7E82P, PJCG8, PJCG9 |
25.5kb | Sense | Chr. 2 - 71055392 - 71056038 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100008312.hg.1 | ENST00000530317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009850.hg.1 | ENST00000505750.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009848.hg.1 | ENST00000509361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008635.hg.1 | ENST00000508347.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300006526.hg.1 | ENST00000383606.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009822.hg.1 | ENST00000425390.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009771.hg.1 | ENST00000454836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008729.hg.1 | ENST00000412208.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012412.hg.1 | ENST00000504655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000309950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000309950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000309950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000309950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000309950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000309950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000309950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000309950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000409042.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000409042.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000409042.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000409042.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000409042.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000409042.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000409042.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000411729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000411729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | NM_001017920.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | NM_001017920.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | NM_001017920.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | NM_001017920.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | NM_001017920.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | NM_001017920.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | NM_001017920.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016553.hg.1 | NM_001017920.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008309.hg.1 | ENST00000532397.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008478.hg.1 | ENST00000448855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006837.hg.1 | ENST00000509439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006837.hg.1 | ENST00000509439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006837.hg.1 | ENST00000509439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006837.hg.1 | ENST00000511021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006837.hg.1 | ENST00000511021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400009838.hg.1 | ENST00000505943.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009825.hg.1 | ENST00000544395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007895.hg.1 | ENST00000448529.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007982.hg.1 | ENST00000400027.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200007982.hg.1 | NR_002185.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007982.hg.1 | NR_002185.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007982.hg.1 | NR_002185.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007982.hg.1 | NR_002185.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200007982.hg.1 | NR_002185.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009670.hg.1 | ENST00000532847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006775.hg.1 | ENST00000504939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006761.hg.1 | ENST00000546439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008633.hg.1 | ENST00000509527.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006773.hg.1 | ENST00000508957.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006773.hg.1 | ENST00000508957.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006773.hg.1 | ENST00000508957.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006773.hg.1 | NM_001040071.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011366.hg.1 | ENST00000533891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008324.hg.1 | ENST00000532736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011580.hg.1 | ENST00000478335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010223.hg.1 | ENST00000496207.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011590.hg.1 | ENST00000494533.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009630.hg.1 | ENST00000528715.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010741.hg.1 | ENST00000605227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009672.hg.1 | ENST00000526420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007421.hg.1 | ENST00000442229.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009876.hg.1 | ENST00000382617.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006834.hg.1 | ENST00000505858.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009624.hg.1 | ENST00000533722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011137.hg.1 | ENST00000502331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011137.hg.1 | ENST00000502331.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011137.hg.1 | ENST00000601375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011137.hg.1 | ENST00000601375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011137.hg.1 | ENST00000601375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008638.hg.1 | ENST00000466578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007151.hg.1 | ENST00000555893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400006838.hg.1 | ENST00000512164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011998.hg.1 | ENST00000489105.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009838.hg.1 | ENST00000505943.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009670.hg.1 | ENST00000532847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011998.hg.1 | ENST00000489105.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009624.hg.1 | ENST00000533722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007895.hg.1 | ENST00000448529.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008309.hg.1 | ENST00000532397.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008312.hg.1 | ENST00000530317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006761.hg.1 | ENST00000546439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007151.hg.1 | ENST00000555893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000309950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000309950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000309950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000309950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000309950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000309950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000309950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000309950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000409042.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000409042.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000409042.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000409042.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000409042.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000409042.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000409042.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000411729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000411729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | ENST00000621326.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | NM_001017920.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | NM_001017920.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | NM_001017920.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | NM_001017920.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | NM_001017920.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | NM_001017920.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | NM_001017920.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016553.hg.1 | NM_001017920.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400006834.hg.1 | ENST00000505858.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009630.hg.1 | ENST00000528715.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007421.hg.1 | ENST00000442229.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007982.hg.1 | ENST00000400027.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200007982.hg.1 | NR_002185.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007982.hg.1 | NR_002185.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007982.hg.1 | NR_002185.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007982.hg.1 | NR_002185.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200007982.hg.1 | NR_002185.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008478.hg.1 | ENST00000448855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011590.hg.1 | ENST00000494533.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009672.hg.1 | ENST00000526420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008729.hg.1 | ENST00000412208.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008633.hg.1 | ENST00000509527.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008635.hg.1 | ENST00000508347.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006775.hg.1 | ENST00000504939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006837.hg.1 | ENST00000509439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006837.hg.1 | ENST00000509439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006837.hg.1 | ENST00000509439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006837.hg.1 | ENST00000511021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006837.hg.1 | ENST00000511021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400009850.hg.1 | ENST00000505750.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010223.hg.1 | ENST00000496207.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006773.hg.1 | ENST00000508957.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006773.hg.1 | ENST00000508957.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006773.hg.1 | ENST00000508957.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006773.hg.1 | NM_001040071.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400009848.hg.1 | ENST00000509361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011137.hg.1 | ENST00000502331.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011137.hg.1 | ENST00000502331.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011137.hg.1 | ENST00000601375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011137.hg.1 | ENST00000601375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011137.hg.1 | ENST00000601375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009876.hg.1 | ENST00000382617.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300006526.hg.1 | ENST00000383606.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008638.hg.1 | ENST00000466578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012412.hg.1 | ENST00000504655.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400006838.hg.1 | ENST00000512164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008324.hg.1 | ENST00000532736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009822.hg.1 | ENST00000425390.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009771.hg.1 | ENST00000454836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011580.hg.1 | ENST00000478335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010741.hg.1 | ENST00000605227.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011366.hg.1 | ENST00000533891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009825.hg.1 | ENST00000544395.1 | Ensembl |
