Assay Details
| Gene Symbol: TSACC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_130913.1 | 2 - 3 | 326 | 109 | ||
| GenBank mRNA | HY048925.1 | - | 2 - 3 | 222 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chaperonin containing TCP1 subunit 3 | CCT3 |
CCT-gamma, CCTG, PIG48, TCP-1-gamma, TRIC5 |
-1.2kb | Anti-sense | Chr. 1 - 156308961 - 156338415 on NCBI Build GRCh38 |
| Downstream | Rh family B glycoprotein (gene/pseudogene) | RHBG | SLC42A2 | 19.0kb | Sense | Chr. 1 - 156366044 - 156385221 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368253.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368253.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368253.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368253.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368254.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000368255.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000466306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000466306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000466306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000466306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000470342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000470342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000470342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000470342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000470342.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000481479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000481479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000481479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000481479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000481479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000481479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | ENST00000481479.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304817.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304817.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304817.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304817.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304817.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304817.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304820.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304820.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304820.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304824.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304824.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304824.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304824.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304824.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304824.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304825.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304825.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304825.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304825.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304825.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304825.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304825.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304826.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304826.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304826.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304826.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_001304826.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_144627.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_144627.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_144627.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_144627.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NM_144627.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NR_130913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NR_130913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NR_130913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NR_130913.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010167.hg.1 | NR_130913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368252.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368253.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368253.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368253.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368253.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368254.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000368255.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000466306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000466306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000466306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000466306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000470342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000470342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000470342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000470342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000470342.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000481479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000481479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000481479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000481479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000481479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000481479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | ENST00000481479.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304817.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304817.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304817.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304817.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304817.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304817.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304820.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304820.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304820.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304824.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304824.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304824.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304824.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304824.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304824.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304825.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304825.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304825.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304825.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304825.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304825.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304825.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304826.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304826.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304826.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304826.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_001304826.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_144627.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_144627.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_144627.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_144627.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NM_144627.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NR_130913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NR_130913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NR_130913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NR_130913.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010167.hg.1 | NR_130913.1 | Refseq |

TSACC
Genomic Map