Assay Details
Gene Symbol: SNORA94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_132776.1 | 1 | 176 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ubiquinol-cytochrome c reductase core protein I | UQCRC1 |
D3S3191, QCR1, UQCR1 |
Sense | Chr. 3 - 48598999 - 48609665 on NCBI Build GRCh38 | |
Downstream | transmembrane protein 89 | TMEM89 | 15.7kb | Sense | Chr. 3 - 48620842 - 48621756 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
