Assay Details
Gene Symbol: MGAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017004147.1 | XP_016859636.1 | 1 - 2 | 367 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017004147.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
