Assay Details
| Gene Symbol: RNF181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016494.3 | NP_057578.1 | 1 - 2 | 139 | 95 | |
| XM_005264359.3 | XP_005264416.1 | 1 - 2 | 585 | 95 | ||
| GenBank mRNA | AF151072.1 | - | 1 - 2 | 119 | 95 | |
| AK222700.1 | - | 1 - 2 | 105 | 95 | ||
| BC002803.2 | - | 1 - 2 | 114 | 3636703 | 95 | |
| CB131242.1 | - | 1 - 2 | 155 | 95 | ||
| CR457165.1 | - | 1 - 2 | 89 | 95 | ||
More Information
Set Membership: |
|

RNF181
Genomic Map