Assay Details
Gene Symbol: SLC20A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017004770.1 | XP_016860259.1 | 5 - 6 | 1164 | 82 | |
XM_017004771.1 | XP_016860260.1 | 6 - 7 | 1200 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017004770.1, XM_017004771.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphate-containing compound metabolic process
transport
ion transport
signal transduction
phosphate ion transmembrane transport
sodium ion transmembrane transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
sodium-dependent phosphate transport
signal transducer activity
receptor activity
inorganic phosphate transmembrane transporter activity
high-affinity inorganic phosphate:sodium symporter activity
sodium:phosphate symporter activity
sodium:inorganic phosphate symporter activity
sodium-dependent phosphate transmembrane transporter activity
phosphate-containing compound metabolic process
transport
ion transport
signal transduction
phosphate ion transmembrane transport
sodium ion transmembrane transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
sodium-dependent phosphate transport
signal transducer activity
receptor activity
inorganic phosphate transmembrane transporter activity
high-affinity inorganic phosphate:sodium symporter activity
sodium:phosphate symporter activity
sodium:inorganic phosphate symporter activity
sodium-dependent phosphate transmembrane transporter activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000492076.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000492076.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000492076.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000492076.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000272542.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000413135.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000423633.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000433924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000456264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000480984.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000490674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000492076.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000492076.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000492076.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000492076.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | ENST00000498224.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008930.hg.1 | NM_005415.4 | Refseq |
