| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000533282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000533282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
ENST00000533282.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000317011.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000531539.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000533282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000533282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
ENST00000533282.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009231.hg.1 |
NM_181721.2 |
Refseq |