Assay Details
| Gene Symbol: NUP50-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038956.1 | 1 - 2 | 204 | 54 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000432502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000432502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000432502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000609284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000609284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000609284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000609284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | ENST00000609432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | NR_038956.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | NR_038956.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | NR_038956.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | NR_038956.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | NR_038956.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | NR_038956.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | NR_038956.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | NR_038956.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | NR_038957.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | NR_038957.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | NR_038957.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | NR_038957.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | NR_038957.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | NR_038957.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008965.hg.1 | NR_038957.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000426282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000432502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000432502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000432502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000609284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000609284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000609284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000609284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | ENST00000609432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | NR_038956.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | NR_038956.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | NR_038956.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | NR_038956.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | NR_038956.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | NR_038956.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | NR_038956.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | NR_038956.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | NR_038957.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | NR_038957.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | NR_038957.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | NR_038957.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | NR_038957.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | NR_038957.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008965.hg.1 | NR_038957.1 | Refseq |

NUP50-AS1
Genomic Map