Assay Details
| Gene Symbol: MYH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011530197.2 | XP_011528499.1 | 2 - 3 | 117 | 116 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011530197.2 transcripts.
Set Membership: |
|

MYH9
Genomic Map