Assay Details
Gene Symbol: PMM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011530232.1 | XP_011528534.1 | 1 - 2 | 113 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011530232.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000493389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000493389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000493389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | ENST00000493389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000216259.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000414636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000463617.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000466645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000472620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000478337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000482178.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000485648.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000493389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000493389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000493389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | ENST00000493389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008831.hg.1 | NM_002676.2 | Refseq |
