Assay Details
| Gene Symbol: DERL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002862.2 | NP_001002862.1 | 7 | 2475 | 67 | |
| NM_198440.3 | NP_940842.2 | 6 | 2576 | 67 | ||
| GenBank mRNA | AK092582.1 | - | 1 | 1620 | 67 | |
| AK096976.1 | - | 6 | 2418 | 67 | ||
| AK125830.1 | - | 5 | 2773 | 67 | ||
| CA313370.1 | - | 1 | 67 | |||
More Information
Set Membership: |
|

DERL3
Genomic Map