Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006869.hg.1 |
ENST00000620093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008196.hg.1 |
ENST00000406213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008196.hg.1 |
ENST00000406213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008196.hg.1 |
ENST00000406213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008196.hg.1 |
ENST00000609510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008196.hg.1 |
ENST00000609510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008196.hg.1 |
ENST00000609510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008196.hg.1 |
ENST00000609510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008196.hg.1 |
NR_038911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008196.hg.1 |
NR_038911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008196.hg.1 |
NR_038911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008196.hg.1 |
NR_038911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006869.hg.1 |
ENST00000620093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008196.hg.1 |
ENST00000406213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008196.hg.1 |
ENST00000406213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008196.hg.1 |
ENST00000406213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008196.hg.1 |
ENST00000609510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008196.hg.1 |
ENST00000609510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008196.hg.1 |
ENST00000609510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008196.hg.1 |
ENST00000609510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008196.hg.1 |
NR_038911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008196.hg.1 |
NR_038911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008196.hg.1 |
NR_038911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008196.hg.1 |
NR_038911.1 |
Refseq |