Assay Details
Gene Symbol: LINC01568 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038234.1 | 2 - 3 | 587 | 92 | ||
GenBank mRNA | BC036646.1 | - | 2 - 3 | 593 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000561875.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000561875.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000561875.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000561875.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000561875.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000561875.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000561875.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000561875.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
