Assay Details
Gene Symbol: SSTR5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC084558.1 | - | 1 | 927 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000565992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000565992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000565992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000565992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000565992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000565992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000565992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000565992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000566499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000566499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000566499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000566499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000566499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569734.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569734.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569734.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569734.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569734.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000624643.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000624643.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | ENST00000624643.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000565992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000565992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000565992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000565992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000565992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000565992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000565992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000565992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000566499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000566499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000566499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000566499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000566499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569734.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569734.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569734.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569734.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569734.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000569832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000624643.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000624643.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | ENST00000624643.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | NR_027242.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009006.hg.1 | NR_027242.1 | Refseq |
