Assay Details
Gene Symbol: BICDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103175.1 | NP_001096645.1 | 2 - 3 | 541 | 81 | |
XM_005255135.4 | XP_005255192.1 | 3 - 4 | 602 | 81 | ||
XM_011522391.1 | XP_011520693.1 | 2 - 3 | 790 | 81 | ||
GenBank mRNA | AK307181.1 | - | 1 - 2 | 85 | 81 | |
AL833717.1 | - | 1 - 2 | 1038 | 81 | ||
AL833749.1 | - | 1 - 2 | 2066 | 81 | ||
BC128602.1 | - | 2 - 3 | 541 | 40132962 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AL833749.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600006655.hg.1 | ENST00000382225.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006655.hg.1 | ENST00000382225.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006655.hg.1 | ENST00000382225.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006655.hg.1 | ENST00000382225.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006655.hg.1 | ENST00000382225.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006655.hg.1 | ENST00000382225.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006655.hg.1 | ENST00000382225.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006655.hg.1 | ENST00000382225.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006655.hg.1 | NR_047572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006655.hg.1 | NR_047572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006655.hg.1 | NR_047572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006655.hg.1 | NR_047572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006655.hg.1 | NR_047572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006655.hg.1 | NR_047572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006655.hg.1 | NR_047572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006655.hg.1 | NR_047572.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000576826.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000576826.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000576826.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000576826.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000576826.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000576826.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000576826.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | ENST00000576826.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000389347.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000572449.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000573514.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000576826.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000576826.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000576826.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000576826.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000576826.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000576826.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000576826.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | ENST00000576826.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009163.hg.1 | NM_001103175.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006655.hg.1 | ENST00000382225.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006655.hg.1 | ENST00000382225.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006655.hg.1 | ENST00000382225.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006655.hg.1 | ENST00000382225.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006655.hg.1 | ENST00000382225.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006655.hg.1 | ENST00000382225.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006655.hg.1 | ENST00000382225.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006655.hg.1 | ENST00000382225.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006655.hg.1 | NR_047572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006655.hg.1 | NR_047572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006655.hg.1 | NR_047572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006655.hg.1 | NR_047572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006655.hg.1 | NR_047572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006655.hg.1 | NR_047572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006655.hg.1 | NR_047572.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006655.hg.1 | NR_047572.1 | Refseq |
