Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
ENST00000338482.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
ENST00000338482.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
ENST00000338482.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
ENST00000338482.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
ENST00000338482.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
ENST00000338482.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
ENST00000338482.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
ENST00000539698.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
ENST00000539698.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
ENST00000539698.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
ENST00000539698.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
ENST00000539698.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
ENST00000539698.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
ENST00000572693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
ENST00000572693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
ENST00000572693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
NM_001166395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
NM_001166395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
NM_001166395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
NM_001166395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
NM_001166395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
NM_001166395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
NM_005769.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
NM_005769.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
NM_005769.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
NM_005769.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008307.hg.1 |
NM_005769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
ENST00000338482.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
ENST00000338482.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
ENST00000338482.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
ENST00000338482.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
ENST00000338482.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
ENST00000338482.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
ENST00000338482.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
ENST00000539698.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
ENST00000539698.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
ENST00000539698.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
ENST00000539698.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
ENST00000539698.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
ENST00000539698.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
ENST00000572693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
ENST00000572693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
ENST00000572693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
NM_001166395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
NM_001166395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
NM_001166395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
NM_001166395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
NM_001166395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
NM_001166395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
NM_005769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
NM_005769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
NM_005769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
NM_005769.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008307.hg.1 |
NM_005769.2 |
Refseq |