Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012199.hg.1 |
ENST00000414755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012199.hg.1 |
ENST00000414755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012199.hg.1 |
ENST00000414755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012199.hg.1 |
ENST00000431149.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012199.hg.1 |
ENST00000431149.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012199.hg.1 |
ENST00000431149.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012199.hg.1 |
ENST00000431149.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012199.hg.1 |
ENST00000431149.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012199.hg.1 |
ENST00000431149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012199.hg.1 |
ENST00000414755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012199.hg.1 |
ENST00000414755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012199.hg.1 |
ENST00000414755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012199.hg.1 |
ENST00000431149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012199.hg.1 |
ENST00000431149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012199.hg.1 |
ENST00000431149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012199.hg.1 |
ENST00000431149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012199.hg.1 |
ENST00000431149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012199.hg.1 |
ENST00000431149.5 |
Ensembl |