Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007543.hg.1 |
ENST00000578602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007543.hg.1 |
ENST00000578602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007543.hg.1 |
ENST00000578602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007543.hg.1 |
ENST00000579896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007543.hg.1 |
ENST00000579896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007543.hg.1 |
ENST00000579896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007543.hg.1 |
ENST00000579896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007543.hg.1 |
ENST00000579896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007543.hg.1 |
ENST00000579896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007543.hg.1 |
ENST00000579975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007543.hg.1 |
ENST00000579975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007543.hg.1 |
ENST00000579975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007543.hg.1 |
ENST00000579975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007543.hg.1 |
ENST00000578602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007543.hg.1 |
ENST00000578602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007543.hg.1 |
ENST00000578602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007543.hg.1 |
ENST00000579896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007543.hg.1 |
ENST00000579896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007543.hg.1 |
ENST00000579896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007543.hg.1 |
ENST00000579896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007543.hg.1 |
ENST00000579896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007543.hg.1 |
ENST00000579896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007543.hg.1 |
ENST00000579975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007543.hg.1 |
ENST00000579975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007543.hg.1 |
ENST00000579975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007543.hg.1 |
ENST00000579975.1 |
Ensembl |