Assay Details
| Gene Symbol: MAPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005257362.4 | XP_005257419.1 | 6 | 693 | 130 | |
| XM_005257364.4 | XP_005257421.1 | 5 | 606 | 130 | ||
| XM_005257365.4 | XP_005257422.1 | 6 | 693 | 130 | ||
| XM_005257366.3 | XP_005257423.1 | 4 | 561 | 130 | ||
| XM_005257367.4 | XP_005257424.1 | 6 | 693 | 130 | ||
| XM_005257368.4 | XP_005257425.1 | 6 | 693 | 130 | ||
| GenBank mRNA | FJ429137.1 | - | 1 | 218 | 130 | |
More Information
Set Membership: |
|

MAPT
Genomic Map