Assay Details
Gene Symbol: LOC100271832 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027097.1 | 4 | 649 | 94 | ||
GenBank mRNA | BC035107.1 | - | 4 | 655 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000363309.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007197.hg.1 | ENST00000363735.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007030.hg.1 | ENST00000365487.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600007333.hg.1 | ENST00000363268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006617.hg.1 | ENST00000384068.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012209.hg.1 | ENST00000384224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009848.hg.1 | ENST00000365638.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500006947.hg.1 | ENST00000384185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009079.hg.1 | ENST00000365176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010078.hg.1 | ENST00000364232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007335.hg.1 | ENST00000365063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009625.hg.1 | ENST00000365381.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011419.hg.1 | ENST00000364757.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010303.hg.1 | ENST00000384695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009842.hg.1 | ENST00000364501.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011735.hg.1 | ENST00000383986.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007642.hg.1 | ENST00000362910.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012209.hg.1 | ENST00000384224.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006617.hg.1 | ENST00000384068.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007333.hg.1 | ENST00000363268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007642.hg.1 | ENST00000362910.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007335.hg.1 | ENST00000365063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007030.hg.1 | ENST00000365487.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000304116.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000355114.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000363309.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000392173.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000585994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000586648.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000587803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | ENST00000591473.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_001042474.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010517.hg.1 | NM_152477.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009079.hg.1 | ENST00000365176.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010078.hg.1 | ENST00000364232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010303.hg.1 | ENST00000384695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009625.hg.1 | ENST00000365381.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011419.hg.1 | ENST00000364757.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007197.hg.1 | ENST00000363735.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011735.hg.1 | ENST00000383986.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500006947.hg.1 | ENST00000384185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009848.hg.1 | ENST00000365638.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009842.hg.1 | ENST00000364501.1 | Ensembl |
