Assay Details
| Gene Symbol: ID2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX095318.1 | - | 1 - 2 | 353 | 66 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000418957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000418957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000418957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000418957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000421298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000421298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000433340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000433340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000433340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000433592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000433592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000433592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000433592.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | NR_110153.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | NR_110153.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000412712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000418957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000418957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000418957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000418957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000421298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000421298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000433340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000433340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000433340.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000433592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000433592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000433592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000433592.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000453900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | ENST00000455965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | NR_110153.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | NR_110153.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | NR_110154.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011664.hg.1 | NR_110154.1 | Refseq |

ID2-AS1
Genomic Map