| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013884.hg.1 |
ENST00000376593.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013884.hg.1 |
ENST00000376593.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013884.hg.1 |
ENST00000376593.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013884.hg.1 |
ENST00000418615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013884.hg.1 |
ENST00000418615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013884.hg.1 |
ENST00000418615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013884.hg.1 |
ENST00000418615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013884.hg.1 |
NR_122074.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013884.hg.1 |
NR_122074.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013884.hg.1 |
NR_122074.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013884.hg.1 |
NR_122074.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013884.hg.1 |
NR_122074.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013884.hg.1 |
ENST00000376593.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013884.hg.1 |
ENST00000376593.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013884.hg.1 |
ENST00000376593.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013884.hg.1 |
ENST00000418615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013884.hg.1 |
ENST00000418615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013884.hg.1 |
ENST00000418615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013884.hg.1 |
ENST00000418615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013884.hg.1 |
NR_122074.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013884.hg.1 |
NR_122074.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013884.hg.1 |
NR_122074.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013884.hg.1 |
NR_122074.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013884.hg.1 |
NR_122074.1 |
Refseq |