Assay Details
| Gene Symbol: CROCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017005450.1 | XP_016860939.1 | 19 - 20 | 3199 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CROCC2| Gene Symbol: CROCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017005450.1 | XP_016860939.1 | 19 - 20 | 3199 | 70 | |
Set Membership: |
|