Assay Details
| Gene Symbol: CROCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017005450.1 | XP_016860939.1 | 20 - 21 | 3319 | 75 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000438506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000438506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000438506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000438506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000438506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000457369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000457369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000457369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000457369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000457369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | ENST00000457369.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000418218.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000438506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000438506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000438506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000438506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000438506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000457369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000457369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000457369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000457369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000457369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | ENST00000457369.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | NR_033841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016339.hg.1 | NR_033841.1 | Refseq |

CROCC2
Genomic Map