| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000413227.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000413227.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000437979.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000437979.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
ENST00000437979.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000413227.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000413227.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000421463.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000437979.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000437979.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
ENST00000437979.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016653.hg.1 |
NR_110259.1 |
Refseq |