Assay Details
Gene Symbol: SNRPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001317165.1 | NP_001304094.1 | 4 - 5 | 411 | 108 | |
GenBank mRNA | BP424287.1 | - | 4 - 5 | 351 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000272348.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000272348.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000272348.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000272348.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000272348.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000272348.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000413456.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000413456.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000413456.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000413456.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000429728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000429728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000429728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000429728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000429728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000429728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000438261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000438261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000438261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000438261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000438261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000449935.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000449935.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000449935.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000449935.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000454893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000454893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000454893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000454893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000454893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000454893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000480370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000480370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000480370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000480370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000480370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000482975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000482975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000482975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000482975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000482975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000482975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000482975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000482975.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000488400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000488400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000488400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | ENST00000488400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | NM_003096.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | NM_003096.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | NM_003096.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | NM_003096.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | NM_003096.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012994.hg.1 | NM_003096.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000272348.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000272348.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000272348.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000272348.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000272348.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000272348.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000413456.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000413456.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000413456.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000413456.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000429728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000429728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000429728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000429728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000429728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000429728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000438261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000438261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000438261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000438261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000438261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000449935.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000449935.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000449935.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000449935.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000454893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000454893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000454893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000454893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000454893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000454893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000480370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000480370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000480370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000480370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000480370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000482975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000482975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000482975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000482975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000482975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000482975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000482975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000482975.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000488400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000488400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000488400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | ENST00000488400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | NM_003096.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | NM_003096.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | NM_003096.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | NM_003096.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | NM_003096.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012994.hg.1 | NM_003096.3 | Refseq |
