Assay Details
Gene Symbol: KCNH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK098545.1 | - | 1 - 2 | 182 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000328032.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000332142.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000477019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000618399.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | ENST00000621889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_033272.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014775.hg.1 | NM_173162.2 | Refseq |
