Assay Details
| Gene Symbol: TRMT61B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005264393.2 | XP_005264450.1 | 5 - 6 | 1316 | 121 | |
| XM_017004401.1 | XP_016859890.1 | 3 - 4 | 1022 | 121 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000419999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000419999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000419999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000419999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000419999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000439947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000439947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000439947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000439947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000439947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000439947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000439947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000484060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000484060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000484060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000490390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000490390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000490390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | ENST00000490390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000306108.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000419999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000419999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000419999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000419999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000419999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000439947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000439947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000439947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000439947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000439947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000439947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000439947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000484060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000484060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000484060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000490390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000490390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000490390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | ENST00000490390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | NM_017910.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012129.hg.1 | NM_017910.3 | Refseq |

TRMT61B
Genomic Map