Assay Details
| Gene Symbol: KANSL1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124963.1 | - | 1 - 2 | 431 | 99 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000429908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000429908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000429908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000460256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000460256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000460256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000460256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |

KANSL1L
Genomic Map