Assay Details
| Gene Symbol: TGFBRAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142621.2 | NP_001136093.1 | 12 | 119 | ||
| NM_004257.5 | NP_004248.2 | 12 | 119 | |||
| XM_017005339.1 | XP_016860828.1 | 11 | 119 | |||
| GenBank mRNA | BC019841.1 | - | 1 | 119 | ||
| BM672953.1 | - | 1 | 217 | 119 | ||
More Information
Set Membership: |
|

TGFBRAP1
Genomic Map