Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000426713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000426713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000426713.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000611969.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
ENST00000611969.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000414416.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000426713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000426713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000426713.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000611969.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
ENST00000611969.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013865.hg.1 |
NR_027063.1 |
Refseq |