Assay Details
Gene Symbol: LINC00116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC133660.1 | - | 4 - 5 | 774 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000426713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000426713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000426713.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000611969.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | ENST00000611969.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000414416.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000426713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000426713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000426713.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000611969.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | ENST00000611969.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | NR_027063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200013865.hg.1 | NR_027063.1 | Refseq |
