Assay Details
Gene Symbol: SPHKAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142644.1 | NP_001136116.1 | 9 - 10 | 4763 | 91 | |
XM_005246870.4 | XP_005246927.1 | 10 - 11 | 5235 | 91 | ||
XM_006712777.3 | XP_006712840.1 | 10 - 11 | 5196 | 91 | ||
XM_011511925.2 | XP_011510227.1 | 11 - 12 | 5314 | 91 | ||
GenBank mRNA | AK095042.1 | - | 3 - 4 | 925 | 91 | |
BC104954.1 | - | 9 - 10 | 4763 | 8143957 | 91 | |
BC104956.1 | - | 9 - 10 | 4763 | 8143959 | 91 | |
CR749297.1 | - | 3 - 4 | 3869 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000490603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000490603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000490603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000490603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | ENST00000490603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000344657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000392056.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000490603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000490603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000490603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000490603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | ENST00000490603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_001142644.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_030623.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015939.hg.1 | NM_030623.3 | Refseq |
