Assay Details
| Gene Symbol: LBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282430.1 | NP_001269359.1 | 1 | 58 | 83 | |
| GenBank mRNA | AY203953.1 | - | 1 | 217 | 83 | |
| BC150517.1 | - | 1 | 42 | 40132747 | 83 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000479098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000479098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000618878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000618878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000618878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000479098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000479098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000618878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000618878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000618878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001282430.1 | Refseq |

LBX2
Genomic Map