Assay Details
| Gene Symbol: LBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC144442.1 | - | 1 - 2 | 386 | 76 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000479098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000479098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000618878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000618878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | ENST00000618878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000341396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000377566.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000460508.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000479098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000479098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000550249.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000618878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000618878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | ENST00000618878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001009812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001282430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013108.hg.1 | NM_001282430.1 | Refseq |

LBX2
Genomic Map