Assay Details
| Gene Symbol: IL18R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282399.1 | NP_001269328.1 | 1 - 2 | 123 | 120 | |
| NM_003855.3 | NP_003846.1 | 1 - 2 | 123 | 120 | ||
| XM_017005182.1 | XP_016860671.1 | 1 - 2 | 588 | 120 | ||
| XM_017005183.1 | XP_016860672.1 | 1 - 2 | 589 | 120 | ||
| XM_017005184.1 | XP_016860673.1 | 1 - 2 | 589 | 120 | ||
| GenBank mRNA | DR006178.1 | - | 2 - 3 | 333 | 120 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001282399.1, NM_003855.3, XM_017005182.1, XM_017005183.1, XM_017005184.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
immune response
signal transduction
natural killer cell activation
positive regulation of interferon-gamma production
interleukin-18-mediated signaling pathway
positive regulation of NF-kappaB import into nucleus
T-helper 1 cell differentiation
receptor activity
interleukin-1 receptor activity
protein binding
interleukin-18 receptor activity
immune response
signal transduction
natural killer cell activation
positive regulation of interferon-gamma production
interleukin-18-mediated signaling pathway
positive regulation of NF-kappaB import into nucleus
T-helper 1 cell differentiation
receptor activity
interleukin-1 receptor activity
protein binding
interleukin-18 receptor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000334376.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000334376.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000334376.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000334376.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000466357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000466357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000466357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000466357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | ENST00000466357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000233957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000334376.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000334376.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000334376.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000334376.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000409599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000466357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000466357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000466357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000466357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | ENST00000466357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_001282399.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_003855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016502.hg.1 | NM_003855.3 | Refseq |

IL18R1
Genomic Map