Assay Details
Gene Symbol: ERICH6-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_121675.1 | 1 - 2 | 112 | |||
GenBank mRNA | BP431953.1 | - | 2 - 3 | 241 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000463755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000463755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000463755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000463755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000463755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000463755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121675.1 | Refseq |
