Assay Details
Gene Symbol: CFAP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006713623.2 | XP_006713686.1 | 1 - 2 | 86 | 70 | |
XM_017006320.1 | XP_016861809.1 | 1 - 2 | 86 | 70 | ||
XM_017006321.1 | XP_016861810.1 | 1 - 2 | 86 | 70 | ||
XM_017006322.1 | XP_016861811.1 | 1 - 2 | 86 | 70 | ||
XM_017006323.1 | XP_016861812.1 | 1 - 2 | 86 | 70 | ||
XM_017006324.1 | XP_016861813.1 | 1 - 2 | 86 | 70 | ||
XM_017006325.1 | XP_016861814.1 | 1 - 2 | 86 | 70 | ||
XM_017006326.1 | XP_016861815.1 | 1 - 2 | 86 | 70 | ||
XM_017006327.1 | XP_016861816.1 | 1 - 2 | 86 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006713623.2, XM_017006320.1, XM_017006321.1, XM_017006322.1, XM_017006323.1, XM_017006324.1, XM_017006325.1, XM_017006326.1, XM_017006327.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000503905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000503905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000503905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000507830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000507830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000507830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000507830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000533201.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000533201.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000503905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000503905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000503905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000507830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000507830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000507830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000507830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000533201.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000533201.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
