Assay Details
Gene Symbol: SPSB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK056147.1 | - | 2 - 3 | 283 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000310546.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000310546.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000310546.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000310546.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000310546.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000310546.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000310546.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000310546.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000507895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000507895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000507895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000507895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000507895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000508126.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000508126.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000508126.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000508126.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000508126.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000508828.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | ENST00000508828.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | NM_080862.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | NM_080862.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | NM_080862.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | NM_080862.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | NM_080862.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | NM_080862.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | NM_080862.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008991.hg.1 | NM_080862.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000310546.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000310546.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000310546.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000310546.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000310546.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000310546.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000310546.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000310546.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000507895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000507895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000507895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000507895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000507895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000508126.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000508126.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000508126.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000508126.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000508126.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000508828.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | ENST00000508828.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | NM_080862.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | NM_080862.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | NM_080862.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | NM_080862.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | NM_080862.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | NM_080862.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | NM_080862.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008991.hg.1 | NM_080862.2 | Refseq |
