Assay Details
Gene Symbol: PRICKLE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011533432.2 | XP_011531734.1 | 1 - 2 | 252 | 57 | |
XM_011533433.2 | XP_011531735.1 | 1 - 2 | 252 | 57 | ||
XM_011533440.2 | XP_011531742.1 | 1 - 2 | 252 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000485770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000485770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000485770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000485770.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000498162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000498162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000498162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000498162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000498162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000498162.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000569824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000569824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000569824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000569824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000569824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | ENST00000569824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007687.hg.1 | ENST00000487097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007687.hg.1 | ENST00000487097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007687.hg.1 | ENST00000487097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007687.hg.1 | ENST00000487097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007687.hg.1 | ENST00000487097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007687.hg.1 | ENST00000487097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007687.hg.1 | ENST00000487097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007687.hg.1 | ENST00000487097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007687.hg.1 | ENST00000487097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007687.hg.1 | ENST00000487097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007687.hg.1 | ENST00000487097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007687.hg.1 | ENST00000487097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007687.hg.1 | ENST00000487097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007687.hg.1 | ENST00000487097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000295902.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000485770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000485770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000485770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000485770.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000498162.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000498162.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000498162.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000498162.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000498162.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000498162.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000564377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000569824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000569824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000569824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000569824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000569824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | ENST00000569824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | NM_198859.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011386.hg.1 | NM_198859.3 | Refseq |
