Assay Details
Gene Symbol: PROS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314077.1 | NP_001301006.1 | 1 - 2 | 403 | 92 | |
GenBank mRNA | BG702968.1 | - | 1 - 2 | 186 | 92 | |
KU178284.1 | - | 1 - 2 | 71 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
platelet degranulation
signal peptide processing
ER to Golgi vesicle-mediated transport
blood coagulation
negative regulation of endopeptidase activity
peptidyl-glutamic acid carboxylation
regulation of complement activation
response to lipopolysaccharide
fibrinolysis
positive regulation of phagocytosis
leukocyte migration
endopeptidase inhibitor activity
calcium ion binding
protein complex binding
platelet degranulation
signal peptide processing
ER to Golgi vesicle-mediated transport
blood coagulation
negative regulation of endopeptidase activity
peptidyl-glutamic acid carboxylation
regulation of complement activation
response to lipopolysaccharide
fibrinolysis
positive regulation of phagocytosis
leukocyte migration
endopeptidase inhibitor activity
calcium ion binding
protein complex binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000472684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000472684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000472684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000472684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000472684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000472684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000488658.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000488658.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | ENST00000488658.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000348974.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000394236.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000407433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000472684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000472684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000472684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000472684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000472684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000472684.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000488658.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000488658.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | ENST00000488658.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011757.hg.1 | NM_000313.3 | Refseq |
