Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
ENST00000340634.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
ENST00000340634.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
ENST00000340634.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
ENST00000340634.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
ENST00000340634.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
ENST00000340634.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
ENST00000492509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
ENST00000492509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
ENST00000492509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
ENST00000492509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
ENST00000498470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
ENST00000498470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
ENST00000498470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
NM_198504.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
NM_198504.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
NM_198504.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
NM_198504.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012679.hg.1 |
NM_198504.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
ENST00000340634.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
ENST00000340634.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
ENST00000340634.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
ENST00000340634.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
ENST00000340634.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
ENST00000340634.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
ENST00000492509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
ENST00000492509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
ENST00000492509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
ENST00000492509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
ENST00000498470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
ENST00000498470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
ENST00000498470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
NM_198504.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
NM_198504.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
NM_198504.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
NM_198504.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012679.hg.1 |
NM_198504.2 |
Refseq |