Assay Details
Gene Symbol: ERICH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152394.4 | NP_689607.2 | 1 - 2 | 470 | 98 | |
XM_005247120.3 | XP_005247177.1 | 1 - 2 | 1062 | 98 | ||
GenBank mRNA | AK093375.1 | - | 2 - 3 | 305 | 98 | |
BC036239.1 | - | 1 - 2 | 454 | 5297417 | 98 | |
DB058602.1 | - | 2 - 3 | 356 | 98 | ||
DB458295.1 | - | 2 - 3 | 350 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000295910.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000474463.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491361.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000491716.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | ENST00000498386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_001308234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012804.hg.1 | NM_152394.4 | Refseq |
