| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009463.hg.1 |
ENST00000404882.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009463.hg.1 |
ENST00000525678.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009463.hg.1 |
ENST00000525678.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009463.hg.1 |
ENST00000525678.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009463.hg.1 |
ENST00000525678.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009463.hg.1 |
ENST00000525678.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009463.hg.1 |
NR_040078.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009463.hg.1 |
NR_040078.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009463.hg.1 |
NR_040078.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009463.hg.1 |
NR_040078.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009463.hg.1 |
NR_040078.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009463.hg.1 |
ENST00000404882.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009463.hg.1 |
ENST00000525678.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009463.hg.1 |
ENST00000525678.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009463.hg.1 |
ENST00000525678.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009463.hg.1 |
ENST00000525678.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009463.hg.1 |
ENST00000525678.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009463.hg.1 |
NR_040078.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009463.hg.1 |
NR_040078.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009463.hg.1 |
NR_040078.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009463.hg.1 |
NR_040078.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009463.hg.1 |
NR_040078.1 |
Refseq |