Assay Details
Gene Symbol: LOC101927204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110184.1 | 2 - 3 | 410 | 98 | ||
GenBank mRNA | AK124475.1 | - | 2 - 3 | 357 | 98 | |
BC028022.1 | - | 2 - 3 | 410 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000533220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000533220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | ENST00000533220.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000501817.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000531708.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000532845.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000533220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000533220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | ENST00000533220.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010959.hg.1 | NR_110184.1 | Refseq |
