Assay Details
Gene Symbol: LOC101928069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120586.1 | 1 - 2 | 211 | 84 | ||
GenBank mRNA | BQ223044.1 | - | 1 - 2 | 193 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000525142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000525142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000525142.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000533502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000533502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000533502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000602951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000602951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | ENST00000602951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | NR_120586.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | NR_120586.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | NR_120586.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | NR_120586.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | NR_120586.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011289.hg.1 | NR_120586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000525142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000525142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000525142.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000527274.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000533502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000533502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000533502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000602951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000602951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | ENST00000602951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | NR_120586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | NR_120586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | NR_120586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | NR_120586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | NR_120586.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011289.hg.1 | NR_120586.1 | Refseq |
