Assay Details
Gene Symbol: GP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005247374.3 | XP_005247431.1 | 1 - 2 | 154 | 99 | |
XM_011512701.1 | XP_011511003.1 | 1 - 2 | 154 | 99 | ||
XM_011512702.1 | XP_011511004.1 | 1 - 2 | 155 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005247374.3, XM_011512701.1, XM_011512702.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
